Dishes, aluminium, flat bottom, with cover
Supplier: Thermo Fisher Scientific
39083.UD
ALFA39083.UDEA 0 SGD
ALFA39083.UD
Dishes, aluminium, flat bottom, with cover
Trays/Dishes/Pots Laboratory Trays/Dishes
Dishes, Aluminium, flat bottom, straight wall, with cover.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Aluminium is the most abundant metallic element on the earth and has many desirable physical and chemical properties. A highly impervious oxide film (approximately 5 nm-thick) is resistant to corrosion by seawater, and other aqueous and chemical solutions. Additionally, this element offers chemical stability in the presence of most organic compounds. Aluminium's reactivity increases with temperature,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable