Bis(pentamethylcyclopentadienyl)nickel(II)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Bis(pentamethylcyclopentadienyl)nickel, Decamethylnickelocene

39238.03 39238.06 39238.MD
ALFA39238.03EA 0 SGD
ALFA39238.03 ALFA39238.06 ALFA39238.MD
Bis(pentamethylcyclopentadienyl)nickel(II)
Bis(pentamethylcyclopentadienyl)nickel (II)

CAS No.: 74507-63-4 Molecular Formula: C20H30Ni Formula Weight: 329.17 Physical Form: Crystalline, Packaged in ampoules Sensitivity: Air and Moisture Sensitive

Formula: C₂₀H₃₀Ni
MW: 329.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058704
CAS Number: 74507-63-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR