Bis(dimethylamino)phosphoryl chloride 94

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Phosphoric Acid Bis(dimethylamido) Chloride, N,N,N',N'-Tetramethylphosphorodiamidic Chloride, Bis(dimethylamino)phosphorochloridate, Bis(dimethylamido)phosphoric chloride

Argon_filled

39274.04 39274.09
ALFA39274.04EA 0 SGD
ALFA39274.04 ALFA39274.09
Bis(dimethylamino)phosphoryl chloride 94
Bis(dimethylamino)phosphoryl chloride

Purity: 94% CAS No.: 1605-65-8 Molecular Formula: C4H12ClN2OP Formula Weight: 170.58 Flash Point: 119°(246°F) Physical Form: Liquid, Packaged under argon Density: 1.152 Refractive Index: 1.4679 Sensitivity: Moisture Sensitive Appearance: Colourless Odour: Pungent Boiling Point: 246-247° Beilstein Reference: 471430 UN No.: UN3265 MDL No.: MFCD00008302

Formula: C₄H₁₂ClN₂OP
MW: 170.58 g/mol
Boiling Pt: 246…247 °C
Density: 1.152
Flash Pt: 119 °C (246 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008302
CAS Number: 1605-65-8
EINECS: 216-517-1
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR