Bis(ethylenedithio)tetrathiafulvalene

Supplier: Thermo Fisher Scientific


Packaged under argon.

39557.MD 39557.03
ALFA39557.MDEA 0 SGD
ALFA39557.MD ALFA39557.03
Bis(ethylenedithio)tetrathiafulvalene
Bis(ethylenedithio)tetrathiafulvalene

CAS No.: 66946-48-3 Molecular Formula: C10H8S8 Formula Weight: 384.68 Physical Form: Powder- Packaged under Argon Melting Point: 238° dec. MDL No.: MFCD00059710

Formula: C₁₀H₈S₈
Melting Pt: 238 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00059710
CAS Number: 66946-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR