pH tester, Checker
Supplier: Thermo Fisher Scientific
39771.KT
ALFA39771.KTEA 0 SGD
ALFA39771.KT
pH tester, Checker
Electrochemistry Meters pH and Multi-Parameter Meters
This pocket size pH meter is supplied with a rugged, epoxy-body pH electrode.
  • Large and easy to read LCD
  • High accuracy with 0,01 pH resolution
  • Two point fast and accurate calibration
  • Secure single action on/off switch
  • Usable with virtually any pH electrode
  • Battery life: 3000 hours
Delivery information: Suplied with screw-type, epoxy-body pH electrode.