Potassium chloride, Ultrapure for spectroscopy

Supplier: Thermo Fisher Scientific
Warning

39795.22
ALFA39795.22EA 0 SGD
ALFA39795.22
Potassium chloride, Ultrapure for spectroscopy
Potassium chloride

CAS No.: 7447-40-7 Molecular Formula: KCl Formula Weight: 74.55 Physical Form: Powder Density: 1.984 Refractive Index: 1.490 Odour: Odourless Melting Point: 773° Boiling Point: 1500° subl. Merck Reference: 14,7621 MDL No.: MFCD00011360

Formula: KCl
Boiling Pt: 1500 °C (subl.)
Melting Pt: 773 °C
Density: 1,984
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,7621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR