Potassium bromide for spectroscopy

Supplier: Thermo Fisher Scientific
Warning

39796.Q4
ALFA39796.Q4EA 0 SGD
ALFA39796.Q4
Potassium bromide for spectroscopy
Potassium bromide

CAS No.: 7758-02-3 Molecular Formula: KBr Formula Weight: 119.01 Physical Form: Random Cuttings, Chunks Density: 2.75 Refractive Index: 1.559 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 730° Boiling Point: 1435° Merck Reference: 14,7618 MDL No.: MFCD00011358

Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435 °C
Melting Pt: 730
Density: 2.75
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 14,7618

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR