Gas analysis crucibles, graphite
Supplier: Thermo Fisher Scientific
39811.KN
ALFA39811.KNEA 0 SGD
ALFA39811.KN
Gas analysis crucibles, graphite
Crucibles
These crucibles are manufactured from graphite.
The crucibles for the analysis of gas content in metals are made from materials exclusively designed for this application. This material, along with a special post treatment of the machined crucible results in a vessel with exceptionally sensitive characteristics for analysing gases in metals.