Tantalum(V) isopropoxide 10% (w/v) in 2-propanol/hexane , 99.9% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

ALFA40038.ACEA 0 SGD
ALFA40038.AC ALFA40038.AE
Tantalum(V) isopropoxide 10% (w/v) in 2-propanol/hexane , 99.9% (metals basis)
Tantalum (V) isopropoxide

Purity: 99.9% CAS No.: 16761-83-4 Molecular Formula: C15H35O5Ta Formula Weight: 476.39 Physical Form: Liquid Sensitivity: Moisture Sensitive UN No.: 1993 MDL No.: MFCD00145347

Formula: C₁₅H₃₅O₅Ta
Storage Temperature: Ambient
MDL Number: MFCD00145347
CAS Number: 16761-83-4
UN: 1993
ADR: 3,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR