Antimony(III) chloride ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Danger

40372.04
ALFA40372.04EA 0 SGD
ALFA40372.04
Antimony(III) chloride ≥99.999% (metals basis), Puratronic®
Antimony (III) chloride

Purity: 99.999% CAS No.: 10025-91-9 Molecular Formula: SbCl3 Formula Weight: 228.11 Physical Form: Crystalline Aggregates Density: 3.14 Melting Point: 73.4° Boiling Point: 223.5° Merck Reference: 14,707 UN No.: 1733 MDL No.: MFCD00011212

Formula: SbCl₃
Boiling Pt: 223,5 °C
Melting Pt: 73,4 °C
Density: 3,14
Storage Temperature: Ambient
MDL Number: MFCD00011212
CAS Number: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8,II
Merck Index: 14,707

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR