Polyethyleneimine, M.W. 1,800 ≥99%, branched

Supplier: Thermo Fisher Scientific
Warning

40528.14 40528.22 40528.36
ALFA40528.14EA 0 SGD
ALFA40528.14 ALFA40528.22 ALFA40528.36
Polyethyleneimine, M.W. 1,800 ≥99%, branched
Polyethyleneimine

Purity: 99% CAS No.: 9002-98-6 Molecular Formula: (-NHCH2CH2-)x(-N(CH2CH2NH2)CH2CH2-)y Physical Form: Liquid Appearance: Colourless to pale yellow Odour: Amine-like MDL No.: MFCD00803910

Formula: (CH₂CH₂NH)n
Storage Temperature: Ambient
MDL Number: MFCD00084427
CAS Number: 9002-98-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR