Polyethyleneimine, M.W. M.W. 70,000 30% (w/v) in aqueous solution, branched

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Polyaziridine, Corcat, Polyethylenimine, Polymin P, Polyaziridine Mw 25000, Aziridine, PEI, Polyethylenimine (branched), Ethyleneimine Polymer, Poly(ethyleneimine)

40529.14 40529.22 40529.36
ALFA40529.14EA 0 SGD
ALFA40529.14 ALFA40529.22 ALFA40529.36
Polyethyleneimine, M.W. M.W. 70,000 30% (w/v) in aqueous solution, branched
Polyethyleneimine

Acts as a protein precipitant used to purify proteins. It is used as chelating agent and as a scavenger for aldehydes and oxides. Also used in detergents, paper industry, dyes, printing inks and in water treatment.


CAS No.: 9002-98-6 Molecular Formula: (-NHCH2CH2-)x(-N(CH2CH2NH2)CH2CH2-)y Physical Form: Liquid Appearance: Colourless to pale yellow Odour: Odourless MDL No.: MFCD00803910


Applications: A protein precipitant used to purify proteins

Formula: (CH₂CH₂NH)n
Storage Temperature: Ambient
MDL Number: MFCD00084427
CAS Number: 9002-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR