Fusion crucibles, graphite
Supplier: Thermo Fisher Scientific
14747.UB 40794.KD
ALFA14747.UBEA 0 SGD
ALFA14747.UB ALFA40794.KD
Fusion crucibles, graphite
Crucibles
These crucibles are manufactured from unpurified extruded graphite.
  • Exceptional thermal uniformity
  • Exactness to dimensional specifications
Order Now
SPECIFICATIONS
Environmentally Preferable