Boats, platinum
Supplier: Thermo Fisher Scientific
41216.KS
ALFA41216.KSEA 0 SGD
ALFA41216.KS
Boats, platinum
Boats Combustion Boats
Combustion boats, Platinum.
  • High melting point
  • High resistance against corrosion
Due to their resistance to oxidation and corrosion, as well as mechanical stability at high temperatures, Platinum boats have become essential for the preparation of samples in analytical laboratories.