Gadolinium(III) isopropoxide ≥99% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

41377.03
ALFA41377.03EA 0 SGD
ALFA41377.03
Gadolinium(III) isopropoxide ≥99% (metals basis)
Gadolinium(III) isopropoxide

Purity: 99% CAS No.: 14532-05-9 Molecular Formula: C9H21GdO3 Formula Weight: 334.51 Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: Off-white Solubility: Soluble in anhydrous 2-propanol, toluene UN No.: UN2925 MDL No.: MFCD00144381


Applications: Sol-Gel synthesis of gadolinium compounds

Formula: C₉H₂₁GdO₃
Storage Temperature: Ambient
MDL Number: MFCD00144381
CAS Number: 14532-05-9
UN: 2925
ADR: 4.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR