DL-Sodium lactate 60% (w/w) in aqueous solution

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (±)-Lactic acid sodium salt, (±)-2-Hydroxypropionic acid sodium salt, Lactate sodium, DL-Lactic acid sodium salt, (±)-2-Hydroxypropanoic acid sodium salt, Sodium 2-hydroxypropanoate

41529.AK 41529.K2
ALFA41529.AKEA 0 SGD
ALFA41529.AK ALFA41529.K2
DL-Sodium lactate 60% (w/w) in aqueous solution
DL-Sodium lactate

CAS No.: 72-17-3 Molecular Formula: CH3CH(OH)CO2Na Formula Weight: 112.06 Physical Form: Liquid Density: 1.326 Appearance: Colourless Odour: Odourless Beilstein Reference: 4332999 Merck Reference: 14,8635 MDL No.: MFCD00065400


Useful chiral synthon; building block for depsipeptides

Formula: H₃CCH(OH)COONa
Density: 1,326
Storage Temperature: Ambient
MDL Number: MFCD00065400
CAS Number: 72-17-3
EINECS: 200-772-0
Merck Index: 14,8635

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR