Chromium(III) chloride hexahydrate ≥98%

Supplier: Thermo Fisher Scientific
Danger

42113.30 42113.A1
ALFA42113.30EA 0 SGD
ALFA42113.30 ALFA42113.A1
Chromium(III) chloride hexahydrate ≥98%
Chromium (III) chloride hexahydrate

Purity: 98% CAS No.: 10060-12-5 Molecular Formula: CrCl3.6H2O Formula Weight: 266.45 (158.35anhy) Physical Form: Crystalline Density: 1.835 Appearance: Dark Green Odour: Odourless Melting Point: 83° Solubility: Slightly soluble in acetone. Soluble in water, alcohol. Insoluble in ether Merck Reference: 14,2222 UN No.: 3260 MDL No.: MFCD00149660

Formula: CrCl₃·6H₂O
Melting Pt: 83 °C
Density: 1,835
Storage Temperature: Ambient
MDL Number: MFCD00149660
CAS Number: 10060-12-5
EINECS: 233-038-3
UN: 3260
ADR: 8,III
Merck Index: 14,2222

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR