Germanium standard solution, 1000 mg/L Ge in water with hydrofluoric acid (max. 1%) (from (NH?)?GeF?), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific

42242.AD 42242.AE 42242.AP
ALFA42242.ADEA 124 SGD
ALFA42242.AD ALFA42242.AE ALFA42242.AP
Germanium standard solution, 1000 mg/L Ge in water with hydrofluoric acid (max. 1%) (from (NH?)?GeF?), standard for ICP, Specpure®
Standards Cationic Standards Germanium Standards

Molecular Formula: (NH₄)₂GeF₆ in H₂O/trace F-
Physical Form: Liquid
MDL No.: MFCD00085310


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR