Fused quartz microscope slides
Supplier: Thermo Fisher Scientific
42295.KG 42295.KJ 42296.KG 42296.KJ 42297.KG 42297.KJ
ALFA42295.KGEA 0 SGD
ALFA42295.KG ALFA42295.KJ ALFA42296.KG ALFA42296.KJ ALFA42297.KG ALFA42297.KJ
Fused quartz microscope slides
Slides Microscope Slides
These fused quartz slides are made of fusing naturally occurring crystalline silica.
  • High chemical purity, high corrosion resistance, high melting point
  • Low coefficient of thermal expansion with excellent electrical insulation
  • Optical transmission from ultra-violet to infrared