Chloramine-T (sodium salt) trihydrate 98.0-103.0% ACS

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt

42374.22 42374.36
ALFA42374.22EA 0 SGD
ALFA42374.22 ALFA42374.36
Chloramine-T (sodium salt) trihydrate 98.0-103.0% ACS
Chloramine T (sodium salt) trihydrate

Purity: 98.0-103.0% CAS No.: 7080-50-4 Molecular Formula: CH3C6H4SO2N(Cl)Na.3H2O Formula Weight: 281.69 (227.67anhy) Flash Point: 192°(377°F) Physical Form: Crystalline Sensitivity: Moisture Sensitive Appearance: White to pale yellow Odour: Chlorine-like pH: 8-10 Melting Point: 167-170° Beilstein Reference: 3924168 Merck Reference: 14,2075 UN No.: 3263 MDL No.: MFCD00149066


Reagent for selective oxidation of methionine

Formula: CH₃C₆H₄SO₂NClNa·3H₂O
MW: 281.69 g/mol
Melting Pt: 167…170 °C
Density: 1.43 g/cm³ (20 °C)
Flash Pt: 192 °C (377 °F)
Storage Temperature: Ambient
MDL Number: MFCD00149066
CAS Number: 7080-50-4
EINECS: 204-854-7
UN: 3263
ADR: 8 III
Merck Index: 14,2075

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR