Sodium nitrite standardised solution 0.1 M in aqueous solution volumetric solution

Supplier: Thermo Fisher Scientific

Synonyms: Diazotizing salts

43015.AP
ALFA43015.APEA 131 SGD
ALFA43015.AP
Sodium nitrite standardised solution 0.1 M in aqueous solution volumetric solution
Sodium nitrite

Formula Weight: 69.00 Physical Form: Liquid Appearance: Colourless Odour: Odourless

Formula: NaNO₂
MW: 69 g/mol
Boiling Pt: 320
Melting Pt: 284
Storage Temperature: Ambient
MDL Number: MFCD00011118
CAS Number: 7632-00-0
EINECS: 231-555-9
UN: 3287
ADR: 6.1 III
Merck Index: 13,08721

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR