Calcium sulphate, Drierite™ 10-20 mesh for drying

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Calcium sulphate (anhydrous) anhydrite, Gypsum

ALFA43058.TDEA 0 SGD
ALFA43058.TD
Calcium sulphate, Drierite™ 10-20 mesh for drying
Calcium sulphate

CAS No.: 7778-18-9 Physical Form: Granular, -10+20 Mesh Density: 2.96 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Merck Reference: 14,1706 MDL No.: MFCD00010912

Formula: CaO₄S
MW: 136.14 g/mol
Melting Pt: 1450 °C
Density: 2.96
Storage Temperature: Ambient
MDL Number: MFCD00010912
CAS Number: 7778-18-9
EINECS: 231-900-3
Merck Index: 14,1706

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR