Methyl cellulose, 1,600 cPs (C=2%, H₂O)

Supplier: Thermo Fisher Scientific

43146.22 43146.36
ALFA43146.22EA 0 SGD
ALFA43146.22 ALFA43146.36
Methyl cellulose, 1,600 cPs (C=2%, H₂O)
Methyl cellulose

CAS No.: 9004-67-5 Physical Form: Powder Odour: Odourless Merck Reference: 14,6040 MDL No.: MFCD00081763

Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS Number: 9004-67-5
Merck Index: 14,6040

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR