Aluminium oxide substrate 12.7x12.7x1 mm, polished one side M plane

Supplier: Thermo Fisher Scientific

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

43246.KT
ALFA43246.KTEA 0 SGD
ALFA43246.KT
Aluminium oxide substrate 12.7x12.7x1 mm, polished one side M plane
Aluminium oxide

Molecular Formula: Al2O3 MDL No.: MFCD00003424

Formula: Al₂O₃
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3,94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR