9,10-Bis(phenylethynyl)anthracene

Supplier: Thermo Fisher Scientific
Warning

43313.03 43313.14 43313.06
ALFA43313.03EA 0 SGD
ALFA43313.03 ALFA43313.14 ALFA43313.06
9,10-Bis(phenylethynyl)anthracene
9,10-Bis(phenylethynyl)anthracene

CAS No.: 10075-85-1 Molecular Formula: C30H18 Formula Weight: 378.47 Physical Form: Crystalline Sensitivity: Light Sensitive Melting Point: 254-255° Beilstein Reference: 1891432 MDL No.: MFCD00012050


As a photochromic material exhibiting chemiluminescence

Formula: C₃₀H₁₈
MW: 378.47 g/mol
Melting Pt: 254…255 °C
Storage Temperature: Ambient
MDL Number: MFCD00012050
CAS Number: 10075-85-1
EINECS: 233-210-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR