Sodium chloride for spectroscopy

Supplier: Thermo Fisher Scientific
Warning

43631.19
ALFA43631.19EA 0 SGD
ALFA43631.19
Sodium chloride for spectroscopy
Sodium chloride

CAS No.: 7647-14-5 Molecular Formula: NaCl Formula Weight: 58.44 Physical Form: Powder Density: 2.165 Refractive Index: 1.5442 Sensitivity: Hygroscopic Appearance: White Odour: Odourless pH: 6-9 Melting Point: 801° Boiling Point: 1413° Merck Reference: 14,8599 MDL No.: MFCD00003477

Formula: NaCl
Boiling Pt: 1413 °C
Melting Pt: 801 °C
Density: 2,165
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,8599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR