Calcium sulphide ≥99% (metals basis excluding Mg) (max. 1% Mg)

Supplier: Thermo Fisher Scientific
Warning

43646.14
ALFA43646.14EA 0 SGD
ALFA43646.14
Calcium sulphide ≥99% (metals basis excluding Mg) (max. 1% Mg)
Calcium sulphide

Purity: 99% CAS No.: 20548-54-3 Molecular Formula: CaS Formula Weight: 72.14 Physical Form: -100 Mesh Powder Density: 2.5 Appearance: White Odour: Stench Melting Point: dec. Merck Reference: 14,1707 UN No.: 3134 MDL No.: MFCD00015990

Formula: CaS
MW: 72.14 g/mol
Melting Pt: Dec.
Density: 2.5
Storage Temperature: Ambient
MDL Number: MFCD00015990
CAS Number: 20548-54-3
EINECS: 243-873-5
UN: 3134
ADR: 4.3 III
Merck Index: 14,1707

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR