Ethyl 2-cyanoacetoacetate

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ethyl2-acetylcyanoacetate

43792.09 43792.04
ALFA43792.09EA 0 SGD
ALFA43792.09 ALFA43792.04
Ethyl 2-cyanoacetoacetate
Ethyl-2-cyanoacetoacetate

CAS No.: 634-55-9 Molecular Formula: C2H5OOCCH(CN)COCH3 Formula Weight: 155.16 Physical Form: Liquid UN No.: UN3276 MDL No.: MFCD00043487

Formula: C₇H₉NO₃
MW: 155.153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043487
CAS Number: 634-55-9
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR