Bis(2,4-pentanedionato)molybdenum(VI) dioxide ≥99%

Supplier: Thermo Fisher Scientific
Warning

44140.09 44140.04 44140.18
ALFA44140.09EA 0 SGD
ALFA44140.09 ALFA44140.04 ALFA44140.18
Bis(2,4-pentanedionato)molybdenum(VI) dioxide ≥99%
Bis(2,4-pentanedionato)molybdenum(VI) dioxide

Purity: 99% CAS No.: 17524-05-9 Molecular Formula: C10H14MoO6 Formula Weight: 326.16 Physical Form: Powder Sensitivity: Air Sensitive Appearance: Pale yellow Melting Point: ca 185° dec. MDL No.: MFCD00011506

Formula: C₁₀H₁₄MoO₆
Melting Pt: ∼ 185 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011506
CAS Number: 17524-05-9
EINECS: 241-522-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR