Multi-element plasma standard solution, non metals, Specpure®

Supplier: Thermo Fisher Scientific

44267.AE 44267.AP
ALFA44267.AEEA 0 SGD
ALFA44267.AE ALFA44267.AP
Multi-element plasma standard solution, non metals, Specpure®
Standards Cationic Standards Multi-Element Standards

As, B, P, S, Se, Si, Te at concentration 100 µg/ml in 5% HCL with a trace of HF.


Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00144997

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR