Bis(cyclopentadienyl)vanadium bromide ≥98%

Supplier: Thermo Fisher Scientific
Danger


ampouled under argon

44360.03
ALFA44360.03EA 0 SGD
ALFA44360.03
Bis(cyclopentadienyl)vanadium bromide ≥98%
Bis(cyclopentadienyl)vanadium bromide

Purity: 98% CAS No.: 64815-29-8 Molecular Formula: C10H10BrV Formula Weight: 261.04 Physical Form: Powder, Ampouled under argon Sensitivity: Air and Moisture Sensitive Boiling Point: 165°/0.2mm subl. UN No.: 3261 MDL No.: MFCD03701567

Formula: C₁₀H₁₀BrV
Boiling Pt: 165 °C (0,2 mmHg) (subl.)
Melting Pt: 221…222 °C
Storage Temperature: Ambient
MDL Number: MFCD03701567
CAS Number: 64815-29-8
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR