Bis(methylcyclopentadienyl)niobium dichloride ≥97%

Supplier: Thermo Fisher Scientific
Danger

44393.03
ALFA44393.03EA 0 SGD
ALFA44393.03
Bis(methylcyclopentadienyl)niobium dichloride ≥97%
Bis(methylcyclopentadienyl)niobium dichloride

Purity: 97% CAS No.: 61374-51-4 Molecular Formula: C12H14Cl2Nb Formula Weight: 322.05 Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: Dark brown Melting Point: 180° dec. UN No.: UN3261 MDL No.: MFCD03427138

Formula: C₁₂H₁₄Cl₂Nb
Melting Pt: 180 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03427138
CAS Number: 61374-51-4
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR