Hexaamminenickel(II) bromide ≥99.999% (metals basis)

Supplier: Thermo Scientific
Danger

44482.14
ALFA44482.14EA 0 SGD
ALFA44482.14
Hexaamminenickel(II) bromide ≥99.999% (metals basis)
Hexaamminenickel (II) bromide

Purity: 99.999% CAS No.: 13601-55-3 Molecular Formula: Ni(NH3)6Br2 Formula Weight: 320.68 Physical Form: Crystalline

Formula: H₁₈Br₂N₆Ni
MW: 320,68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039935
CAS Number: 13601-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR