Phenol ≥99.5% unstabilised

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

44526.22
ALFA44526.22EA 0 SGD
ALFA44526.22
Phenol ≥99.5% unstabilised
Phenol

Unstabilized


Purity: 99.5% CAS No.: 108-95-2 Molecular Formula: C6H5OH Formula Weight: 94.11 Flash Point: 79°(175°F) Physical Form: Solid melt Density: 1.071 Sensitivity: Hygroscopic Odour: Sweet Melting Point: 40-43° Boiling Point: 181° Beilstein Reference: 969616 Merck Reference: 14,7241 UN No.: 1671 MDL No.: MFCD00002143


Molecular biology

Formula: C₆H₅OH
MW: 94.11 g/mol
Boiling Pt: 181 °C
Melting Pt: 40…43 °C
Density: 1.071
Flash Pt: 79 °C (175 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1 II
REACH: 01-2119471329-32
Merck Index: 14,7241

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR