Lithium chloride, dry_ultra 99,995 Metals basis

Supplier: Thermo Fisher Scientific
Warning


ampuled under argon

44762.14 44762.22
ALFA44762.14EA 0 SGD
ALFA44762.14 ALFA44762.22
Lithium chloride, dry_ultra 99,995 Metals basis
Lithium chloride

Purity: 99.995% CAS No.: 7447-41-8 Molecular Formula: LiCl Formula Weight: 42.39 Physical Form: Powder, Ampouled under argon Density: 2.068 Melting Point: 605° Boiling Point: 1325-1360° Merck Reference: 14,5528 MDL No.: MFCD00011078

Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1325…1360 °C
Melting Pt: 605 °C
Density: 2.068
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 14,5528

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR