2-Bromo-3-(2-tetrahydropyranyloxy)pyridine ≥97%

Supplier: Thermo Fisher Scientific
Warning

44774.06
ALFA44774.06EA 0 SGD
ALFA44774.06
2-Bromo-3-(2-tetrahydropyranyloxy)pyridine ≥97%
2-Bromo-3-(2-tetrahydropyranyloxy)pyridine

Purity: 97% CAS No.: 93560-60-2 Molecular Formula: C10H12BrNO2 Formula Weight: 258.12 Physical Form: Solid Melting Point: 67° MDL No.: MFCD06411700

Formula: C₁₀H₁₂BrNO₂
MW: 258.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06411700
CAS Number: 93560-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR