Methyl cellulose, 15 cPs

Supplier: Thermo Fisher Scientific

45490.A4 45490.22 45490.36
ALFA45490.A4EA 0 SGD
ALFA45490.A4 ALFA45490.22 ALFA45490.36
Methyl cellulose, 15 cPs
Methyl cellulose

CAS No.: 9004-67-5 Physical Form: Powder Odour: Odourless MDL No.: MFCD00081763

Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS Number: 9004-67-5
Merck Index: 14,6040

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR