Polyethyleneimine, M.W. 1,200 ≥99%, branched

Supplier: Thermo Fisher Scientific
Warning

45594.14 45594.22 45594.36
ALFA45594.14EA 0 SGD
ALFA45594.14 ALFA45594.22 ALFA45594.36
Polyethyleneimine, M.W. 1,200 ≥99%, branched
Polyethyleneimine

Purity: 99% CAS No.: 9002-98-6 Molecular Formula: (-NHCH2CH2-)x(-N(CH2CH2NH2)CH2CH2-)y Physical Form: Liquid Appearance: Pale yellow Odour: Amine-like MDL No.: MFCD00803910

Formula: (CH2CH2NH)n
Storage Temperature: Ambient
MDL Number: MFCD00084427
CAS Number: 9002-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR