Ion exchange resins, Diaion® HP20
Supplier: Thermo Fisher Scientific
46488.A1
ALFA46488.A1EA 288 SGD
ALFA46488.A1
Ion exchange resins, Diaion® HP20
Chromatography Sorbents HPLC Sorbents
Diaion® HP20 is used as a filler in chromatography columns due to its extremely large pores. It is considered as an effective adsorbent to eliminate toxic effects in fermentation and separation of traces heavy metal ions. It is also useful for the purification of peptides, proteins, fermentation products, polyphenols and cephalosporin C. It helps to remove bitter tasting substances from fruit juices.
Order Now
SPECIFICATIONS
Environmentally Preferable