Dibutyltin bis(2-ethylhexanoate)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Di-n-butyltin bis(2-ethylhexanoate), Dibutylstannanediyl bis(2-ethylhexanoate)

71126.06 71126.14 71126.22
ALFA71126.06EA 0 SGD
ALFA71126.06 ALFA71126.14 ALFA71126.22
Dibutyltin bis(2-ethylhexanoate)
Dibutyltin bis(2-ethylhexanoate)

CAS No.: 2781-10-4 Molecular Formula: C24H48O4Sn Formula Weight: 519.34 Physical Form: Lump Density: 1.070 Odour: Odourless Boiling Point: 220°/2mm Solubility: Soluble in mineral oil, acetone, xylene UN No.: 3146 MDL No.: MFCD00015261

Formula: C₂₄H₄₈O₄Sn
MW: 519.34 g/mol
Boiling Pt: 220 °C (2 mmHg)
Melting Pt: 57…59 °C
Density: 1.070
Storage Temperature: Ambient
MDL Number: MFCD00015261
CAS Number: 2781-10-4
EINECS: 220-481-2
UN: 3146
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR