Diboron trioxide ≥99.98% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Boron oxide, Boric anhydride

89964.36 89964.22
ALFA89964.36EA 0 SGD
ALFA89964.36 ALFA89964.22
Diboron trioxide ≥99.98% (metals basis)
di-Boron trioxide

Purity: 99.98% CAS No.: 1303-86-2 Molecular Formula: B2O3 Formula Weight: 69.62 Physical Form: Powder Density: 2.46 Appearance: White Odour: Odourless Melting Point: ca 450° Boiling Point: 1860° Merck Reference: 14,1337 MDL No.: MFCD00011315

Formula: B₂O₃
MW: 69.62 g/mol
Boiling Pt: 1860 °C
Melting Pt: ∼450 °C
Density: 2.46
Storage Temperature: Ambient
MDL Number: MFCD00011315
CAS Number: 1303-86-2
EINECS: 215-125-8
REACH: 01-2119486655-24
Merck Index: 14,1337

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR