Glove bags, small
Supplier: Thermo Fisher Scientific
91717.LK 91717.LL
ALFA91717.LKEA 0 SGD
ALFA91717.LK ALFA91717.LL
Glove bags, small
Glove Boxes and Glove Bags
These polyethylene glove bags are ideal for laboratory use and field sampling.
Suitable for handling air and moisture sensitive, high purity chemicals and materials. Soluble in xylene.
Delivery information: Supplied with closure, tubing coupler, and cotton gloves.