Glove bags, large
Supplier: Thermo Fisher Scientific
92717.LK 92717.LL 92727.LK 92727.LL 93727.LK 93727.LL 93737.LK 93737.LL
ALFA92717.LKEA 0 SGD
ALFA92717.LK ALFA92717.LL ALFA92727.LK ALFA92727.LL ALFA93727.LK ALFA93727.LL ALFA93737.LK ALFA93737.LL
Glove bags, large
Glove Boxes and Glove Bags
These polyethylene glove bags are used for handling air and moisture sensitive chemicals in laboratories. Also suitable for field sampling.
  • Minimise work exposure
  • Preserve the integrity of the sample during storage
  • Serve as a portable laboratory for conducting tests and crime scene investigations
Delivery information: Supplied with closure, tubing coupler, and cotton gloves.
Order Now
SPECIFICATIONS
Environmentally Preferable