Ethyl salicylate ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Ethyl 2-hydroxybenzoate

A10101.0E A10101.22 A10101.36
ALFAA10101.0EEA 0 SGD
ALFAA10101.0E ALFAA10101.22 ALFAA10101.36
Ethyl salicylate ≥99%
Ethyl salicylate

Purity: 99% CAS No.: 118-61-6 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 107°(224°F) Refractive Index: 1.5220 Melting Point: 1° Boiling Point: 232° Beilstein Reference: 907659 Merck Reference: 14,3850 MDL No.: MFCD00002215

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 232 °C
Melting Pt: 1
Density: 1.130
Flash Pt: 107 °C (224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002215
CAS Number: 118-61-6
EINECS: 204-265-5
Merck Index: 13,03881

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR