2-Chlorophenol ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Chloro-2-hydroxybenzene

A10311.30 A10311.0B A10311.0I
ALFAA10311.30EA 0 SGD
ALFAA10311.30 ALFAA10311.0B ALFAA10311.0I
2-Chlorophenol ≥99%
2-Chlorophenol

Purity: 99% CAS No.: 95-57-8 Molecular Formula: C6H5ClO Formula Weight: 128.56 Flash Point: 63°(145°F) Refractive Index: 1.5590 Odour: Phenol-like Melting Point: 7-8° Boiling Point: 175-176° Beilstein Reference: 1905114 Merck Reference: 14,2154 UN No.: 2021 MDL No.: MFCD00002159

Formula: C₆H₅ClO
Boiling Pt: 175…176 °C
Melting Pt: 7…8 °C
Density: 1,260
Flash Pt: 63 °C (145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002159
CAS Number: 95-57-8
EINECS: 202-433-2
UN: 2021
ADR: 6.1,III
Merck Index: 14,2154

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR