4,4'-Isopropylidenediphenol ≥97%

Supplier: Thermo Fisher Scientific
Danger

A10324.0B A10324.0I A10324.30
ALFAA10324.0BEA 0 SGD
ALFAA10324.0B ALFAA10324.0I ALFAA10324.30
4,4'-Isopropylidenediphenol ≥97%
4,4'-Isopropylidenediphenol

Purity: 97+% CAS No.: 80-05-7 Molecular Formula: C15H16O2 Formula Weight: 228.29 Flash Point: 227°(440°F) Melting Point: 153-158° Boiling Point: 200°/4mm Beilstein Reference: 1107700 Merck Reference: 14,1297 MDL No.: MFCD00002366

Formula: C₁₅H₁₆O₂
Boiling Pt: 200 °C (4 mmHg)
Melting Pt: 153…158 °C
Density: 1,205 g/cm³ (20 °C)
Flash Pt: 227 °C (440 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002366
CAS Number: 80-05-7
EINECS: 201-245-8
Merck Index: 14,1297

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR