4-Nitrocinnamyl alcohol 98%

Supplier: Thermo Fisher Scientific
Warning

A10338.03 A10338.06 A10338.14
ALFAA10338.03EA 0 SGD
ALFAA10338.03 ALFAA10338.06 ALFAA10338.14
4-Nitrocinnamyl alcohol 98%
4-Nitrocinnamyl alcohol

Purity: 98% CAS No.: 1504-63-8 Molecular Formula: C9H9NO3 Formula Weight: 179.18 Melting Point: 126-130° Beilstein Reference: 1948238 MDL No.: MFCD00017045

Formula: C₉H₉NO₃
MW: 179.18 g/mol
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00017045
CAS Number: 1504-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR