Trityl alcohol ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Triphenylmethanol, Triphenylcarbinol

A10366.0B A10366.18 A10366.30
ALFAA10366.0BEA 0 SGD
ALFAA10366.0B ALFAA10366.18 ALFAA10366.30
Trityl alcohol ≥98%
Trityl alcohol

Purity: 98% CAS No.: 76-84-6 Molecular Formula: (C6H5)3COH Formula Weight: 260.34 Physical Form: Cryst powder Appearance: White to cream Melting Point: 161-164° Boiling Point: 360-380° Beilstein Reference: 1460837 Merck Reference: 14,9739 MDL No.: MFCD00004445

Formula: C₁₉H₁₆O
MW: 260.34 g/mol
Boiling Pt: 360…380 °C
Melting Pt: 161…164 °C
Density: 1.1884 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004445
CAS Number: 76-84-6
EINECS: 200-988-5
Merck Index: 13,09810

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR