Sodium gluconate ≥97%

Supplier: Thermo Fisher Scientific

Synonyms: D-Gluconic acid sodium salt, Sodium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic acid, sodium salt, Sodium D-gluconate

A10464.0C A10464.0E
ALFAA10464.0CEA 0 SGD
ALFAA10464.0C ALFAA10464.0E
Sodium gluconate ≥97%
Sodium gluconate

Purity: 97% CAS No.: 527-07-1 Molecular Formula: C6H11NaO7 Formula Weight: 218.14 Physical Form: Cryst powder Appearance: White Odour: Pleasant Melting Point: ca 206° dec. Beilstein Reference: 3919651 Merck Reference: 14,4456 Optical Rotation: +12° (c=20 in water) MDL No.: MFCD00064210

Formula: HOH₂C(CH(OH))₄COONa
Melting Pt: ∼ 206 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064210
CAS Number: 527-07-1
EINECS: 208-407-7
Merck Index: 14,4456
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eafd5b786cb23fb5c20dd64ad056ed2d80dc0186&vid=b1ffe48dafca4b8bd9b0d709ea3e986099051767&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR