Bismuth(III) chloride, Wasser_3 98 Dry weight

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Bismuth(III) chloride, Bismuth trichloride

A10510.14 A10510.22 A10510.36
ALFAA10510.14EA 0 SGD
ALFAA10510.14 ALFAA10510.22 ALFAA10510.36
Bismuth(III) chloride, Wasser_3 98 Dry weight
Bismuth (III) chloride

Purity: 98% CAS No.: 7787-60-2 Molecular Formula: BiCl3 Formula Weight: 315.34 Odour: Irritating Melting Point: 230-232° Boiling Point: ca 430° subl. Merck Reference: 14,1262 UN No.: 3260 MDL No.: MFCD00003461

Formula: BiCl₃
MW: 315.34 g/mol
Boiling Pt: ∼430 °C (subl.)
Melting Pt: 230…232 °C
Density: 4.75
Storage Temperature: Ambient
MDL Number: MFCD00003461
CAS Number: 7787-60-2
EINECS: 232-123-2
UN: 3260
ADR: 8 III
Merck Index: 14,1262

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR